missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p55 gamma Antibody (2F8), Novus Biologicals™

Product Code. 18389689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389689 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389689 Supplier Novus Biologicals Supplier No. H00008503M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI 3-Kinase p55 gamma Monoclonal antibody specifically detects PI 3-Kinase p55 gamma in Human samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase p55 gamma
Applications Western Blot, ELISA, Proximity Ligation Assay, Sandwich ELISA
Classification Monoclonal
Clone 2F8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH21622
Gene Alias DKFZp686P05226, FLJ41892, p55, p55-GAMMA, p55PIK, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphatidylinositol 3-kinase regulatory subunit gamma, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit 3 (gamma), PI3K regulatory subunit gamma, PI3-kinase regulatory subunit gamma, PI3-kinase subunit p55-gamma, PIK3R3, PtdIns-3-kinase regulatory subunit gamma, ptdIns-3-kinase regulatory subunit p55-gamma, regulatory subunit, polypeptide 3 (p55, gamma)
Host Species Mouse
Immunogen PIK3R3 (AAH21622, 271 a.a. ~ 380 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HDSKMRLEQDLKNQALDNREIDKKMNSIKPDLIQLRKIRDQHLVWLNHKGVRQKRLNVWLGIKNEDAAENYFINEEDENLPHYDEKTWFVEDINRVQAEDLLYGKPDGAF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, mTOR Pathway, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8503
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.