Learn More
Eurofins DiscoverX PI 3-Kinase (p110α(H1047R)/p85α) Human Protein
Description
The H1047R substitution is a somatic mutation in p110α that has been associated with tumours of the colon, stomach, breast and brain. Combined in vitro and in vivo studies have shown that this mutation confers higher lipid kinase activity than wild type, and is able to induce oncogenic transformation.
GenBank U79143 / GenBank XM_043865
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 129 kDa(p110α(H1047R)) and 83.6 kDa (p85α) |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / Untagged |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | p110a ≥36 & p85a ≥34 Combined ≥70% |
| Protein | PI 3-Kinase (p110α(H1047R)/p85α) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.