missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3-Kinase p110 gamma Antibody (OTI2B1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46411
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
PI 3-Kinase p110 gamma Monoclonal specifically detects PI 3-Kinase p110 gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PI 3-Kinase p110 gamma | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:150 | |
| 1-phosphatidylinositol 3-kinase, EC 2.7.1, EC 2.7.1.153, p110-gamma, p120-PI3K, phosphatidylinositol 3 kinase gamma, p110 gamma, phosphatidylinositol 3-kinase catalytic 110-kD gamma, phosphatidylinositol 3-kinase, catalytic, gamma polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform, phosphoinositide-3-kinase gamma catalytic subunit, phosphoinositide-3-kinase, catalytic, gamma polypeptide, PI3CG, PI3K, PI3Kgamma, PI3K-gamma, PI3-kinase subunit gamma, PIK3, PtdIns-3-kinase subunit gamma, PtdIns-3-kinase subunit p110-gamma | |
| Mouse | |
| 126.3 kDa | |
| 0.1 mL | |
| mTOR Pathway, Signal Transduction | |
| 5294 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI2B1 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| PIK3CG | |
| Full length human recombinant protein of human PIK3CG(NP_002640) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction