missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p110 delta Rabbit anti-Human, Mouse, Rat, Clone: 3J0F6, Novus Biologicals™

Product Code. 18348225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348225 20 μg 20µL
18305654 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18348225 Supplier Novus Biologicals Supplier No. NBP31588920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PI 3-Kinase p110 delta Monoclonal antibody specifically detects PI 3-Kinase p110 delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase p110 delta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3J0F6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.7.1, EC 2.7.1.153, p110D, P110DELTA, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform, phosphoinositide-3-kinase C, phosphoinositide-3-kinase, catalytic, delta polypeptide, PI3K, PI3K-delta, PI3-kinase p110 subunit delta, PI3-kinase subunit delta, PtdIns-3-kinase subunit delta, PtdIns-3-kinase subunit p110-delta
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human PI 3-Kinase p110 delta (O00329).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Autophagy, mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5293
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.