missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3-Kinase p110 delta Antibody (PIK3CD/4639), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24232MFV450
This item is not returnable.
View return policy
Description
PI 3-Kinase p110 delta Monoclonal antibody specifically detects PI 3-Kinase p110 delta in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| PI 3-Kinase p110 delta | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| EC 2.7.1, EC 2.7.1.153, p110D, P110DELTA, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform, phosphoinositide-3-kinase C, phosphoinositide-3-kinase, catalytic, delta polypeptide, PI3K, PI3K-delta, PI3-kinase p110 subunit delta, PI3-kinase subunit delta, PtdIns-3-kinase subunit delta, PtdIns-3-kinase subunit p110-delta | |
| Recombinant fragment of human PI 3-Kinase p110 delta protein (around aa 520-720) (exact sequence is proprietary) | |
| 0.1 mL | |
| Autophagy, mTOR Pathway, Phospho Specific, Signal Transduction | |
| 5293 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| PIK3CD/4639 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction