Learn More
PI 3-Kinase p110 beta/PIK3CB Antibody (CL9518) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88937DL594
Product Code. 30292579
  • £558.00 / 0.10mL
Description
PI 3-Kinase p110 beta/PIK3CB Monoclonal antibody specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PI 3-Kinase p110 beta/PIK3CB
Monoclonal
DyLight 594
P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta
This antibody was developed using a synthetic peptide derived from P42338, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Cancer, Cell Cycle and Replication, mTOR Pathway
5291
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL9518
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified