missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p110 beta/PIK3CB Antibody (10D5), Novus Biologicals™

Product Code. 18420110 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420110 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420110 Supplier Novus Biologicals Supplier No. H00005291M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI 3-Kinase p110 beta/PIK3CB Monoclonal antibody specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase p110 beta/PIK3CB
Applications Western Blot, ELISA, Immunocytochemistry, Proximity Ligation Assay
Classification Monoclonal
Clone 10D5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006210
Gene Alias P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta
Host Species Mouse
Immunogen PIK3CB (NP_006210, 147 a.a. ~ 256 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EFRRKMRKFSEEKILSLVGLSWMDWLKQTYPPEHEPSIPENLEDKLYGGKLIVAVHFENCQDVFSFQVSPNMNPIKVNELAIQKRLTIHGKEDEVSPYDYVLQVSGRVEY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5291
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.