missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase C2 beta Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30735974 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735974 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735974 Supplier Novus Biologicals Supplier No. AF7249MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PI 3-Kinase C2 beta Polyclonal antibody specifically detects PI 3-Kinase C2 beta in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase C2 beta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias C2-PI3KDKFZp686G16234, EC 2.7.1, EC 2.7.1.154, phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta, Phosphoinositide 3-kinase-C2-beta, phosphoinositide-3-kinase, class 2, beta polypeptide, PI3K-C2beta, PI3K-C2-beta, PTDINS-3-kinase C2 beta, PtdIns-3-kinase C2 subunit beta
Host Species Goat
Immunogen E. coli-derived recombinant human PI 3-Kinase C2β, Ser2-Ser134, Accession # O00750
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5287
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.