missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PI(3,4)P2 Lipid Kinase Substrate

Product Code. 17108835
Click to view available options
Quantity:
400 nmol
5000 nmol
Unit Size:
400nmol
5000nmol
This item is not returnable. View return policy

Product Code. 17108835

Brand: Invitrogen™ A33597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C)

  • The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C).
  • The substrate consists of solubilized phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.
  • Validated for use with Thermo Fisher Scientific lipid kinases
  • Available in multiple pack sizes for convenient use
  • Also available in our >SelectScreen™ Kinase Profiling Service
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in 30 mM HEPES pH 7.5 buffer.
Form Powder
Product Line Invitrogen
Quantity 400 nmol
Substrate Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2)
Substrate Type Kinase Substrate
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.