missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI(3,4)P2 Lipid Kinase Substrate
Click to view available options
Quantity:
400 nmol
5000 nmol
Unit Size:
400nmol
5000nmol
Description
- The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C)
- The substrate consists of solubilized phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in an aqueous buffer
- It can be used in combination with an Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors
- Validated for use with Thermo Fisher Scientific lipid kinases
- Available in multiple pack sizes for convenient use
- Also available in our SelectScreen™ Kinase Profiling Service
Specifications
Specifications
| Content And Storage | Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in 30 mM HEPES pH 7.5 buffer. |
| Form | Powder |
| Product Line | Invitrogen |
| Quantity | 5000 nmol |
| Substrate | Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) |
| Substrate Type | Kinase Substrate |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu