missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI(3,4)P2 Lipid Kinase Substrate
Click to view available options
Quantity:
400 nmol
5000 nmol
Unit Size:
400nmol
5000nmol
Description
- The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C)
- The substrate consists of solubilized phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in an aqueous buffer
- It can be used in combination with an Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors
- Validated for use with Thermo Fisher Scientific lipid kinases
- Available in multiple pack sizes for convenient use
- Also available in our SelectScreen™ Kinase Profiling Service
Specifications
Specifications
| Content And Storage | Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in 30 mM HEPES pH 7.5 buffer. |
| Form | Powder |
| Product Line | Invitrogen™ |
| Quantity | 5000 nmol |
| Substrate | Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) |
| Substrate Type | Kinase Substrate |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction