missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Phytone™ Peptone

Product Code. 16229881
Change view
Click to view available options
Quantity:
454 g
10 kg
Unit Size:
10kg
454g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16229881 10 kg 10kg
16269761 454 g 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16229881 Supplier Gibco™ Supplier No. 292450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An animal origin-free (AOF) enzymatic digest of soy.

  • An animal origin-free (AOF) enzymatic digest of soy
  • A nutritious and excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
  • Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics
  • Also used with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines

Applications of Phytone Peptone include:

  • It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
  • It has been used in mammalian cell culture applications due to its high carbohydrate content
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Peptone
For Use With (Application) Production of monoclonal antibodies for oncological and rheumatological therapeutics, microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines
Product Line Phytone
Quantity 10 kg
Source Animal origin-free, soy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.