missing translation for 'onlineSavingsMsg'
Learn More

Phytic Acid Dodecasodium Salt Hydrate, TRC

Product Code. 30414061
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414061 5 g -
30408948 1 g 1g
30414060 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414061 Supplier Toronto Research Chemicals Supplier No. TRCP3987135G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123408-98-0
Molecular Formula C6H6Na12O24P6 . xH2O
Molecular Weight (g/mol) 941.83
Synonym Hexakis(Dihydrogen Phosphate) myo-Inositol Dodecasodium Salt Hydrate
IUPAC Name dodecasodium;(2,3,4,5,6-pentaphosphonatooxycyclohexyl) phosphate;hydrate
SMILES O.[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P(=O)([O-])O[C@@H]1[C@H](OP(=O)([O-])[O-])[C@@H](OP(=O)([O-])[O-])[C@H](OP(=O)([O-])[O-])[C@@H](OP(=O)([O-])[O-])[C@@H]1OP(=O)([O-])[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H18O24P6.12Na.H2O/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15;;;;;;;;;;;;;/h1-6H,(H2,7,8,9)(H2,10,11,12)(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24);;;;;;;;;;;;;1H2/q;12*+1;/p-12/t1-,2-,3-,4+,5-,6-;;;;;;;;;;;;;
Formula Weight 941.6553
Chemical Name or Material Phytic Acid Dodecasodium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.