missing translation for 'onlineSavingsMsg'
Learn More

Phytic acid dodecasodium hydrate, MedChemExpress

Product Code. 16429905
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16429905 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16429905 Supplier Medchem Express Supplier No. HYN0814A
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phytic acid (Inositol hexaphosphate) dodecasodium hydrate is a phosphorus storage compound of seeds and cereal grains.

Phytic acid dodecasodium hydrate is known as a food inhibitor, which has a strong ability to chelate multivalent metal ions, specially zinc, calcium, iron and as with protein residue. Phytic acid dodecasodium hydrate inhibits the enzymatic superoxide source xanthine oxidase (XO), and has antioxidative, neuroprotective, anti-inflammatory effects.

Specifications

Chemical Name or Material Phytic acid dodecasodium hydrate
CAS 123408-98-0
Color White
Purity Grade Notes Research
Molecular Formula C6H18O24P6.XH2O.12Na
Quantity 250 mg
Synonym Inositol hexaphosphate dodecasodium hydrate
Solubility Information H2O : ≥ 250 mg/mL
SMILES O=P(O[Na])(O[Na])O[C@H]1[C@H]([C@H]([C@H](OP(O[Na])(O[Na])=O)[C@@H](OP(O[Na])(O[Na])=O)[C@@H]1OP(O[Na])(O[Na])=O)OP(O[Na])(O[Na])=O)OP(O[Na])(O[Na])=O.[x].O
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.