missing translation for 'onlineSavingsMsg'
Learn More

Innovating Science™ Physical Properties of Glass

Product Code. 13163160
Click to view available options
:
Physical Properties of Glass Kit
This item is not returnable. View return policy

Product Code. 13163160

Brand: Innovating Science™ IS9005

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Teacher's manual and student guide, analysis copy-masters, one bottle of borosilicate glass beads, one bottle of flint glass beads, one bottle of soda-lime glass beads, one set of refractive index solutions, 15 magnifiers. Enough materials for 15 tests.

Requires

Balance, graduated cylinders (10mL), test tubes

See how forensic scientists use the chemical and physical properties of glass to discover evidence.

Police and crime scene investigators must use all available tools and evidence, working backwards to create the most likely scenario as to what might have occurred. Different types of evidence provide different pieces to the puzzle.

Specifications

Color White
Product Type Physical Properties of Glass Kit
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensic scientists use the chemical and physical properties of glass to discover evidence
Includes Teacher's manual and student guide, analysis copy-masters, one bottle of borosilicate glass beads, one bottle of flint glass beads, one bottle of soda-lime glass beads, one set of refractive index solutions, 15 magnifiers, enough materials for 15 tests
Quantity 1
Requires Balance, graduated cylinders (10mL), test tubes
Dimensions (L x W x H) 9.75 x 6.75 x 3 in.
Class Size 15 Lab Groups
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.