missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic XG Safety Gloves with Aqua-Polymer Xtra Grip Foam Coating
GREENER_CHOICE

Product Code. 17260381 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17260381 12 10 pairs
15376912 6 Each
15396912 8 Each
15306922 9 Each
15356912 10 Each
15366912 11 Each
15386912 7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17260381 Supplier uvex Supplier No. 6007012
Add to basket
Add to basket
This item is not returnable. View return policy

Uvex™ Phynomic series gloves are ideal for types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The Xtra Grip aqua-polymer foam coating enhances grip in oily conditions.

  • Xtra Grip aqua-polymer foam coating is breathable and provides a high level of mechanical abrasion resistance
  • Enhanced grip capabilities in oily areas
  • Tactile feel when assembling oily parts
  • Ideal for precision work, maintenance, repair work, and metal processing
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Construction Seamless Knit
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Black, Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Spandex, Polyamide
Product Type Mechanical Protection Gloves
Size 12
Style Full-hand Mitt
EN388 Standard 4121X
For Use With (Application) Assembly, Maintenance, and Precision Work
Gender Unisex
Glove Cuff Style Knit
Series Phynomic XG
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.