missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic Lite Gray Safety Gloves

Product Code. 17200351 Shop All Uvex Products
Click to view available options
Size:
5
6
7
8
9
10
11
12
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17200351

Brand: uvex 6004009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight gloves are constructed from a combination of polyamide and elastane, with aqua polymer impregnation on the palm and fingertips for increased durability and dexterity.

  • Extremely lightweight construction makes these gloves suitable for particularly delicate tasks
  • Aqua-polymer impregnation on the palm and fingertips for mechanical abrasion resistance
  • Enhances grip capabilities in dry and slightly damp areas
  • Porous coating is breathable to reduce perspiration
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
  • 3D Ergo technology ensures a secure fit
  • Suitable for use with food handling and processing
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer
Color Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Elastane, Polyamide
Product Type Mechanical Protection Gloves
Size 9
EN388 Standard 2121X
For Use With (Application) Driving/Operating Machinery, Painting, and Paper Hanging
Gender Unisex
Glove Cuff Style Knit
Series Phynomic Lite
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.