missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic All-Round Safety Gloves

Product Code. 17280361 Shop All Uvex Products
Click to view available options
Size:
5
6
7
8
9
10
11
12
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17280361

Brand: uvex 6004912

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, dirt-resistant gloves are constructed from a blend of polyamide and elastane, with aqua-polymer foam coating on the palm and fingertips to repel moisture.

  • Lightweight construction acts as a second-skin to maintain a high level of feeling
  • Aqua-polymer foam coating is moisture repellent and flexible
  • Breathable materials help prevent excessive perspiration
  • Dirt-resistant exterior is ideal for mechanical tasks
  • Enhanced grip in dry and slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Gray, Black
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 12
EN388 Standard 3131X
For Use With (Application) Assembly, Maintanence, Shipping/Logistics, Precision Work, and Repairs
Gender Unisex
Glove Cuff Style Knit
Series Phynomic All-Round
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.