missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHYKPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584606
This item is not returnable.
View return policy
Description
Immunogen sequence: VLNVLEKEQL QDHATSVGSF LMQLLGQQKI KHPIVGDVRG V Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 73%.
This is a nuclear gene encoding a mitochondrial enzyme that catalyzes the conversion of 5-phosphonooxy-L-lysine to ammonia, inorganic phosphate, and 2-aminoadipate semialdehyde. Mutations in this gene may cause phosphohydroxylysinuria. Alternative splicing results in multiple transcript variants.
Specifications
| PHYKPL | |
| Polyclonal | |
| Unconjugated | |
| PHYKPL | |
| 2900006B13Rik; 5-phosphohydroxy-L-lysine phospholyase; 5-phosphohydroxy-L-lysine phospho-lyase; 5-phosphonooxy-L-lysine phospho-lyase; Agxt2l2; Alanine--glyoxylate aminotransferase 2-like 2; alanine-glyoxylate aminotransferase 2-like 2; PHLU; PHYKPL; PP9286 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUZ5 | |
| PHYKPL | |
| Recombinant protein corresponding to Human PHYKPL. Recombinant protein control fragment (Product #RP-107262). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction