missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYHIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£285.00 - £423.00
Specifications
| Antigen | PHYHIP |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PHYHIP Polyclonal specifically detects PHYHIP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PHYHIP | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DYRK1A interacting protein 3, DYRK1AP3, KIAA0273PAHX-AP1, PAHX-AP, PAHXAP1, phytanoyl-CoA 2-hydroxylase interacting protein, phytanoyl-CoA alpha-hydroxylase associated protein, phytanoyl-CoA hydroxylase interacting protein, Phytanoyl-CoA hydroxylase-associated protein 1, phytanoyl-CoA hydroxylase-interacting protein | |
| PHYHIP | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 9796 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CDSFRISWAMEDSDLERVTHY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title