missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12858-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody (12858-1-AP) detects two bands: 36 kDa and 70 kDa. The 70 kDa band may probably be the dimeric form of PHYH.
PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.Specifications
| PHYH | |
| Polyclonal | |
| Unconjugated | |
| PHYH | |
| LN1, LNAP1, PAHX, PHYH, PHYH1, Phytanic acid oxidase, phytanoyl CoA 2 hydroxylase, RD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114209, 16922, 5264 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14832, O35386, P57093 | |
| Phyh | |
| PHYH Fusion Protein Ag3569 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction