missing translation for 'onlineSavingsMsg'
Learn More

PHYH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12858-1-AP-150UL

 View more versions of this product

Product Code. 16800804

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody (12858-1-AP) detects two bands: 36 kDa and 70 kDa. The 70 kDa band may probably be the dimeric form of PHYH.

PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
Specifications

Specifications

PHYH
Polyclonal
Unconjugated
PHYH
LN1, LNAP1, PAHX, PHYH, PHYH1, Phytanic acid oxidase, phytanoyl CoA 2 hydroxylase, RD
Rabbit
Antigen Affinity Chromatography
RUO
114209, 16922, 5264
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14832, O35386, P57093
Phyh
PHYH Fusion Protein Ag3569
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.