missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ Flash High-Fidelity PCR Master Mix Promotion

Product Code. 15737516 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 reactions
500 reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15737516 100 reactions 100 reactions
15727516 500 reactions 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15737516 Supplier Thermo Scientific™ Supplier No. F548S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Perform short PCR protocols without compromising fidelity or yield with this master mix.

The optimal annealing temperature for Phusion DNA Polymerases may differ significantly from that of Taq-based polymerases.

For optimal results start by accurately calculating your Tm with our Tm calculator.

Phusion Flash PCR Master Mix was developed to save valuable laboratory time. Ready-to-use 2X master mix enables the use of extremely short PCR protocols without compromising either the fidelity or the yield in the reaction. In addition, pipetting steps are minimized: only template and primers need to be added by the user.

The unique composition of the master mix enables the use of extremely short cycling protocols with both low and high complexity DNA templates - 15 seconds per kilobase or less. The master mix utilizes Phusion Flash II DNA Polymerase, a proofreading DNA polymerase modified from Phusion Hot Start II High-Fidelity DNA Polymerase.

Highlights

  • Extreme speed: extension times of 15 s/kb or less
  • Hot start modification allowing “zero-time reactivation”
  • Accuracy: proofreading DNA polymerase with a fidelity of 25X Taq
  • High yields in reduced time
TRUSTED_SUSTAINABILITY

Specifica

Content And Storage Recommended storage temperature -20°C. Stable for six months from the packaging date.
Quantity 100 reactions
For Use With (Application) Cloning

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppels. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.