missing translation for 'onlineSavingsMsg'
Learn More

Phthalylsulfathiazole, TRC

Product Code. 30135028
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135028 10 g 10g
30133162 5 g 5g
30011563 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135028 Supplier Toronto Research Chemicals Supplier No. TRCP39750310G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-73-4
Molecular Formula C17 H13 N3 O5 S2
Molecular Weight (g/mol) 403.43
Synonym Phthalylsulfathiazole, 2-[[4-(Thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid, Benzoic acid, 2-[[[4-[(2-thiazolylamino)sulfonyl]phenyl]amino]carbonyl]-, Phthalanilic acid, 4'-(2-thiazolylsulfamoyl)- (8CI), (o-Carboxybenzoyl)-p-aminophenylsulfonamidothiazole, 2-(N4-Phthalylaminobenzenesulfonamide)thiazole, 2-(N4-Phthalylsulfanilamido)thiazole, 4'-(2-Thiazolylsulfamoyl)phthalanilic acid, 4'-(2-Thiazolylsulfamyl)phthalanilic acid, AFI-Ftalyl, Cremothalidine, Enteramida, Entero-sulfina, Entexidina, Ftalazol, Ftalil-Esteve, Ftalil-Septol, Ftalysept, Intestiazol, NSC 66454, NSC 683525, Phtalazol, Phthalazol, Phthalidin, Phthalylnorsulfazole, Phthalylsulfathiazole, Phthalylsulfonazole, Phthalylsulphathiazole, Sulfacetil, Sulfaphthalazole, Sulfathalidine, Sulftalyl, Sulphaphthalyl, Taleudron, Talidine, Taloudron, Thalazole, Thalinil, Thalistanin, Thalistatyl, Ultratiazol
IUPAC Name 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid
SMILES OC(=O)c1ccccc1C(=O)Nc2ccc(cc2)S(=O)(=O)Nc3nccs3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H13N3O5S2/c21-15(13-3-1-2-4-14(13)16(22)23)19-11-5-7-12(8-6-11)27(24,25)20-17-18-9-10-26-17/h1-10H,(H,18,20)(H,19,21)(H,22,23)
Formula Weight 403.0297
Chemical Name or Material N4-Phthalylsulfathiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.