missing translation for 'onlineSavingsMsg'
Learn More

Phthalylsulfathiazole, Mikromol™

Product Code. 17998844
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17998844 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17998844 Supplier Mikromol Supplier No. MM1935.000100
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phthalylsulfathiazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N4-Phthalylsulfathiazole
CAS 85-73-4
Analyte or Component Names N4-Phthalylsulfathiazole
Shipping Condition Room Temperature
Molecular Formula C17 H13 N3 O5 S2
Certifications/Compliance ISO 17034
Synonym Phthalylsulfathiazole, 2-[[4-(Thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid, Benzoic acid, 2-[[[4-[(2-thiazolylamino)sulfonyl]phenyl]amino]carbonyl]-, Phthalanilic acid, 4'-(2-thiazolylsulfamoyl)- (8CI), (o-Carboxybenzoyl)-p-aminophenylsulfonamidothiazole, 2-(N4-Phthalylaminobenzenesulfonamide)thiazole, 2-(N4-Phthalylsulfanilamido)thiazole, 4'-(2-Thiazolylsulfamoyl)phthalanilic acid, 4'-(2-Thiazolylsulfamyl)phthalanilic acid, AFI-Ftalyl, Cremothalidine, Enteramida, Entero-sulfina, Entexidina, Ftalazol, Ftalil-Esteve, Ftalil-Septol, Ftalysept, Intestiazol, NSC 66454, NSC 683525, Phtalazol, Phthalazol, Phthalidin, Phthalylnorsulfazole, Phthalylsulfathiazole, Phthalylsulfonazole, Phthalylsulphathiazole, Sulfacetil, Sulfaphthalazole, Sulfathalidine, Sulftalyl, Sulphaphthalyl, Taleudron, Talidine, Taloudron, Thalazole, Thalinil, Thalistanin, Thalistatyl, Ultratiazol
InChI Formula InChI=1S/C17H13N3O5S2/c21-15(13-3-1-2-4-14(13)16(22)23)19-11-5-7-12(8-6-11)27(24,25)20-17-18-9-10-26-17/h1-10H,(H,18,20)(H,19,21)(H,22,23)
SMILES OC(=O)c1ccccc1C(=O)Nc2ccc(cc2)S(=O)(=O)Nc3nccs3
IUPAC Name 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid
Molecular Weight (g/mol) 403.43
Quantity 100 mg
Formula Weight 403.0297 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.