missing translation for 'onlineSavingsMsg'
Learn More

Phthalylsulfathiazole, MedChemExpress

Product Code. 16422276
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422276 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422276 Supplier Medchem Express Supplier No. HYB1407
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Phthalylsulfathiazole is a kind of sulfonamides used as an antibacterial drug.

Phthalylsulfathiazole is a kind of sulfonamides used as an antibacterial drug.

Specifications

Chemical Name or Material Phthalylsulfathiazole
Physical Form Solid
CAS 85-73-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C17H13N3O5S2
Synonym N4-Phthalylsulfathiazole
Solubility Information DMSO : ≥ 40 mg/mL (99.15 mM)
SMILES O=C(O)C1=CC=CC=C1C(NC2=CC=C(S(=O)(NC3=NC=CS3)=O)C=C2)=O
Molecular Weight (g/mol) 403.43
Formula Weight 403.43
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.