missing translation for 'onlineSavingsMsg'
Learn More

Phthalocyanine

Product Code. 11398317
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11398317 25 g 25g
11388317 1 g 1g
11378317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11398317 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer 032073.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 574-93-6 | C32H18N8 | 514.552 g/mol

Pthalocyanine is a common macrocylic blue-green dye. It is also used as a suitable laser dye metal. Phthalocyanines have long been examined as catalysts for redox reactions. Areas of interest are the oxygen reduction reaction and the sweetening of gas streams by removal of hydrogen sulfide. Phthalocyanine compounds have been investigated as donor materials in molecular electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 574-93-6
Molecular Formula C32H18N8
Molecular Weight (g/mol) 514.552
MDL Number MFCD00005085
InChI Key IEQIEDJGQAUEQZ-UHFFFAOYSA-N
Synonym phthalocyanine, pigment blue 16, 29h,31h-phthalocyanine, phthalocyanin, heliogen blue g, polymon blue g, lionol blue kw, irgazin blue 3gt, ftalocianina, ci pigment blue 16
PubChem CID 5282330
ChEBI CHEBI:34921
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3

Specifikationer

Odor Odorless
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 514.52
Physical Form Crystalline
Chemical Name or Material Phthalocyanine

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.