missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, TRC

Product Code. 30424361
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424361 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424361 Supplier Toronto Research Chemicals Supplier No. TRCP384550500G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalimide
CAS 85-41-6
Molecular Formula C8 H5 N O2
Quantity 500 g
Synonym Phthalimide, 1H-Isoindole-1,3(2H)-dione, Phthalimide (8CI), 1,2-Benzenedicarboximide, 1,3-Dihydro-2H-isoindole-1,3-dione, 1,3-Dioxo-1,3-dihydroisoindole, 1,3-Isoindolinedione, 2,3-Dihydro-1H-isoindole-1,3-dione, Benzoimide, Isoindole-1,3-dione, Kladnoite, Levegal PEW-T, NSC 3108, Phenylimide, Phthalic dicarboximide
InChI Formula InChI=1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-4H,(H,9,10,11)
SMILES O=C1NC(=O)c2ccccc12
IUPAC Name isoindole-1,3-dione
Molecular Weight (g/mol) 147.13
Formula Weight 147.032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.