missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, Potassium salt, 99%, MP Biomedicals™

Product Code. 11488290 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488290 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11488290 Supplier MP Biomedicals Supplier No. 0210264980
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1074-82-4
Color Yellow
Assay Percent Range 99%
Molecular Formula C8H4KNO2
MDL Number MFCD00005887
Quantity 100 g
Synonym potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
InChI Key FYRHIOVKTDQVFC-UHFFFAOYSA-M
SMILES [K+].O=C1[N-]C(=O)C2=CC=CC=C12
IUPAC Name potassium 1,3-dioxo-2,3-dihydro-1H-isoindol-2-ide
Molecular Weight (g/mol) 185.22
PubChem CID 3356745
Formula Weight 185.2
Physical Form Powder
Chemical Name or Material Phthalimide, Potassium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.