missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, potassium derivative, 99%, Thermo Scientific Chemicals

Catalog Number p-4510545
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
1074-82-4
C8H4KNO2
185.22
MFCD00005887
FYRHIOVKTDQVFC-UHFFFAOYSA-M
potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
3356745
potassium;isoindol-2-ide-1,3-dione
[K+].O=C1[N-]C(=O)C2=CC=CC=C12
1074-82-4
C8H4KNO2
185.22
MFCD00005887
FYRHIOVKTDQVFC-UHFFFAOYSA-M
potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
3356745
potassium;isoindol-2-ide-1,3-dione
[K+].O=C1[N-]C(=O)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color Green or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Solubility Information Solubility in water: soluble in water
Formula Weight 185.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Phthalimide, potassium derivative
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'