missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, 99%

Product Code. 11494294
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494294 2500 g 2500g
11484294 500 g 500g
11404304 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494294 Supplier Thermo Scientific Alfa Aesar Supplier No. A12584.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85-41-6 | C8H5NO2 | 147.13 g/mol

Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.

Solubility
Slightly soluble in water. Soluble in aqueous alkali hydroxides

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-41-6
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00005881
InChI Key XKJCHHZQLQNZHY-UHFFFAOYSA-N
Synonym phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
PubChem CID 6809
ChEBI CHEBI:38817
IUPAC Name isoindole-1,3-dione
SMILES O=C1NC(=O)C2=CC=CC=C12

Specifications

Melting Point 232°C to 236°C
Density 1.21
pH 3.8
Boiling Point 366°C
Flash Point 165°C (329°F)
Odor Odorless
Quantity 2500 g
Beilstein 118522
Merck Index 14,7373
Solubility Information Slightly soluble in water; Soluble in aqueous alkali hydroxides
Formula Weight 147.14
Percent Purity 99%
Chemical Name or Material Phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.