missing translation for 'onlineSavingsMsg'
Learn More

Phthalide, TRC

Product Code. 30423410
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423410 50 g 50g
30420715 100 g -
30424636 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423410 Supplier Toronto Research Chemicals Supplier No. TRCP38452550G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-41-2
Molecular Formula C8 H6 O2
Molecular Weight (g/mol) 134.13
Synonym 1(3H)-Isobenzofuranone (9CI, ACI), Phthalide (6CI, 8CI), 1,3-Dihydro-2-benzofuran-1-one, 1-Phthalanone, 2-Benzofuran-1(3H)-one, 2-Hydroxymethylbenzoic acid γ-lactone, 3H-Isobenzofuran-1-one, NSC 1469, Phthalolactone
IUPAC Name 3H-2-benzofuran-1-one
SMILES O=C1OCc2ccccc12

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H6O2/c9-8-7-4-2-1-3-6(7)5-10-8/h1-4H,5H2
Formula Weight 134.0368
Chemical Name or Material Isobenzofuranone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.