missing translation for 'onlineSavingsMsg'
Learn More

Phthalic anhydride, 99%

Produktkod. 11497147
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1000 g
5000 g
Förpackningsstorlek:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
11497147 1000 g 1000g
11417157 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11497147 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A14955.0B
ChemsChrom Mega Deals
Use promo code 27993
Visa alla kampanjer
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol

Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.

Solubility
Soluble in water, carbon disulfide, ethanol, acetone and benzene. Slightly soluble in ethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
MDL Number MFCD00005918
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
PubChem CID 6811
ChEBI CHEBI:36605
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifikationer

Melting Point 130°C to 134°C
Density 1.53
Boiling Point 295°C (sublimation)
Flash Point 152°C (305°F)
Odor Weak, Characteristic
Quantity 1000 g
UN Number UN2214
Beilstein 118515
Sensitivity Moisture sensitive
Merck Index 14,7372
Solubility Information Soluble in water,carbon disulfide,ethanol,acetone and benzene. Slightly soluble in ethyl ether.
IUPAC Name 2-benzofuran-1,3-dione
Formula Weight 148.12
Percent Purity 99%
Chemical Name or Material Phthalic anhydride
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.