missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10626462 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10626462 500 g 500g
10636462 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626462 Supplier Fisher Chemical Supplier No. P/3360/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
MDL Number MFCD00005918
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
PubChem CID 6811
ChEBI CHEBI:36605
IUPAC Name 2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

Packaging Plastic powder jar
Vapor Pressure 0.01mbar at 20°C
Physical Form Solid
Color White
pH 2
Melting Point 129°C
Boiling Point 284°C
Quantity 500 g
Formula Weight 148.12g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phthalic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.