missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, 99%

Product Code. 10113350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113350 2.5 kg Plastic bottle 2.5kg
10428803 250 g Plastic bottle 250g
10655782 1 kg Plastic bottle 1kg
11905051 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10113350 Supplier Thermo Scientific Acros Supplier No. 154960025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
MDL Number MFCD00005918
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
PubChem CID 6811
ChEBI CHEBI:36605
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

Melting Point 129.0°C to 132.0°C
Boiling Point 284.0°C
Flash Point 152°C
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 17,469
Fieser 01,882; 17,148
Merck Index 15,7484
Solubility Information Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water
IUPAC Name 1,3-dihydro-2-benzofuran-1,3-dione
Viscosity 6.4 mPa.s (197°C)
Formula Weight 148.12
Percent Purity 99%
Chemical Name or Material Phthalic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.