missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, n-pentyl-isopentyl ester (mixture of isomers), Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Dipentyl phthalate (mix of isomers) |
| CAS | 84777-06-0 |
| Molecular Formula | C18 H26 O4 |
| Quantity | 100 mg |
| Synonym | 1,2-Benzenedicarboxylic Acid, Dipentylester, Branched and Linear, 1,2-Benzenedicarboxylic acid, dipentyl ester, branched and linear, Dipentyl phthalate branched and linear, Phthalic acid, n-pentyl-isopentyl ester (mixture of isomers), Phthalic Acid, n-pentyl-isopentyl Ester (mixture of isomers) |
| Molecular Weight (g/mol) | 306.4 |
| Formula Weight | 306.18 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?