missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-n-pentyl ester, Dr. Ehrenstorfer

Product Code. 30318038
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30318038 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318038 Supplier Dr. Ehrenstorfer Supplier No. DREC16175500
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-n-pentyl ester
CAS 131-18-0
Molecular Formula C18 H26 O4
Quantity 250 mg
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dipentyl ester, 1,2-Benzenedicarboxylic acid, dipentyl ester (9CI), Phthalic acid, dipentyl ester (6CI,7CI,8CI), 1,2-Dipentyl benzene-1,2-dicarboxylate, Amoil, Amyl phthalate, Di-n-amyl phthalate, Di-n-pentyl o-phthalate, Di-n-pentyl phthalate, Diamyl phthalate, Dipentyl phthalate, NSC 4720, Phthalic acid dipentyl ester, Phthalic acid, bis-n-pentyl ester
InChI Formula InChI=1S/C18H26O4/c1-3-5-9-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-10-6-4-2/h7-8,11-12H,3-6,9-10,13-14H2,1-2H3
SMILES CCCCCOC(=O)c1ccccc1C(=O)OCCCCC
IUPAC Name dipentyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 306.4
Formula Weight 306.1831 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.