missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-n-heptyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315168
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315168 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315168 Supplier Dr. Ehrenstorfer Supplier No. DREA16173100AL100
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid,bis-n-heptyl ester
CAS 3648-21-3
Molecular Formula C22 H34 O4
Quantity 1 mL
InChI Formula InChI=1S/C22H34O4/c1-3-5-7-9-13-17-25-21(23)19-15-11-12-16-20(19)22(24)26-18-14-10-8-6-4-2/h11-12,15-16H,3-10,13-14,17-18H2,1-2H3
SMILES CCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCC
IUPAC Name diheptyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 362.5
Formula Weight 362.2457 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.