missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-methylglycol ester, Dr. Ehrenstorfer

Product Code. 30326059
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30326059 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326059 Supplier Dr. Ehrenstorfer Supplier No. DREC16174400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-methylglycol ester
CAS 117-82-8
Molecular Formula C14 H18 O6
Quantity 250 mg
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-methoxyethyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-methoxyethyl) ester (9CI), Phthalic acid, bis(2-methoxyethyl) ester (6CI,7CI,8CI), Ethanol, 2-methoxy-, phthalate (2:1) (8CI), 2-Methoxyethyl phthalate, Bis(2-methoxyethyl) phthalate, Bis(2-methoxyethyl)ester 1,2-benzenedicarboxylic acid, Bis(methoxyethyl) phthalate, Di(2-methoxyethyl) phthalate, Dimethyl glycol phthalate, KC 988, Kesscoflex MCP, Methyl glycol phthalate, NSC 2147
InChI Formula InChI=1S/C14H18O6/c1-17-7-9-19-13(15)11-5-3-4-6-12(11)14(16)20-10-8-18-2/h3-6H,7-10H2,1-2H3
SMILES COCCOC(=O)c1ccccc1C(=O)OCCOC
IUPAC Name bis(2-methoxyethyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 282.29
Formula Weight 282.1103 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.