missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-isobutyl ester, Dr. Ehrenstorfer

Product Code. 30321052
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 30321052

Brand: Dr. Ehrenstorfer DREC16173500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-iso-butyl ester
CAS 84-69-5
Molecular Formula C16 H22 O4
Quantity 100 mg
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-methylpropyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-methylpropyl) ester (9CI), Phthalic acid, diisobutyl ester (6CI,7CI,8CI), 1,2-Benzenedicarboxylic acid diisobutyl ester, 2-Methylpropyl phthalate, Bis(2-methylpropyl) o-phthalate, Bis(2-methylpropyl) phthalate, Di(2-methylpropyl) phthalate, Di(isobutyl) 1,2-benzenedicarboxylate, Di-iso-Butyl phthalate, Diisobutyl phthalate, Hexaplas M/1B, Isobutyl phthalate, NSC 15316, Palatinol IC, Reomol DiBP, Phthalic acid, bis-isobutyl ester
InChI Formula InChI=1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3
SMILES CC(C)COC(=O)c1ccccc1C(=O)OCC(C)C
IUPAC Name bis(2-methylpropyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 278.34
Formula Weight 278.1518 g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.