missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, bis-hexyl ester 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-hexyl ester |
| CAS | 84-75-3 |
| Molecular Formula | C20 H30 O4 |
| Quantity | 10 mL |
| Synonym | 1,2-Benzenedicarboxylic acid, 1,2-dihexyl ester, 1,2-Benzenedicarboxylic acid, dihexyl ester (9CI), Phthalic acid, dihexyl ester (6CI,7CI,8CI), Bis(n-hexyl) phthalate, Di-n-hexyl phthalate, Dihexyl phthalate, Jayflex DHP, NSC 4817, Phthalic acid dihexyl ester, Phthalic acid, bis-hexyl ester |
| InChI Formula | InChI=1S/C20H30O4/c1-3-5-7-11-15-23-19(21)17-13-9-10-14-18(17)20(22)24-16-12-8-6-4-2/h9-10,13-14H,3-8,11-12,15-16H2,1-2H3 |
| SMILES | CCCCCCOC(=O)c1ccccc1C(=O)OCCCCCC |
| IUPAC Name | dihexyl benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 334.45 |
| Formula Weight | 334.2144 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?