missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, ACS, 99.5+%

Product Code. p-7051685
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348918 100 g 100g
11338918 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348918 Supplier Thermo Scientific Alfa Aesar Supplier No. 041770.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-99-3 | C8H6O4 | 166.132 g/mol

Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-99-3
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002467
InChI Key XNGIFLGASWRNHJ-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
PubChem CID 1017
ChEBI CHEBI:29069
IUPAC Name phthalic acid
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 205°C (decomposition)
Assay Percent Range >99.5%
Beilstein 608199
Merck Index 14,7371
Solubility Information Soluble in alcohol. Slightly soluble in water,methanol and ether. Insoluble in chloroform.
Formula Weight 166.13
Percent Purity ≥99.5%
Grade ACS
Density 1.593
Quantity 100 g
Flash Point 168°C (334°F)
Physical Form Crystalline Powder
Chemical Name or Material Phthalic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.