missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, 99%

Product Code. p-3776550 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10745781

Brand: Thermo Scientific Acros 131072500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-99-3 | C8H6O4 | 166.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-99-3
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002467
InChI Key XNGIFLGASWRNHJ-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
PubChem CID 1017
ChEBI CHEBI:29069
IUPAC Name phthalic acid
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 200.0°C to 205.0°C
Color White
Assay Percent Range 99%
Beilstein 09, 791
Merck Index 15, 7483
Solubility Information Solubility in water: 7g/L (25°C). Other solubilities: soluble in alcohol, sparingly soluble in ether, practically insoluble in chloroform
Formula Weight 166.13
Percent Purity 99%
Density 168g/mL
Quantity 250 g
Flash Point 168°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Fine Crystalline Powder
Chemical Name or Material Phthalic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phthalic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.