missing translation for 'onlineSavingsMsg'
Learn More

Phthalhydrazide, 98%

Product Code. 11412058
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412058 250 g 250g
11434030 10 g 10g
11402058 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412058 Supplier Thermo Scientific Alfa Aesar Supplier No. A14615.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1445-69-8 | C8H6N2O2 | 162.15 g/mol

Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

Solubility
Soluble in acetone and acetic acid.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-69-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00006888
InChI Key JNQUJPBFSQETPT-UHFFFAOYNA-N
Synonym 2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
PubChem CID 219401
SMILES O=C1N=NC(=O)C2C=CC=CC12

Specifications

Melting Point 342°C to 345°C
Quantity 250 g
Beilstein 135926
Solubility Information Soluble in acetone and acetic acid.
IUPAC Name 1,4,4a,8a-tetrahydrophthalazine-1,4-dione
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Phthalhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.