missing translation for 'onlineSavingsMsg'
Learn More

Phthalazone, TRC

Product Code. 30427923
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427923 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427923 Supplier Toronto Research Chemicals Supplier No. TRCP382600500MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalazone
CAS 119-39-1
Purity Grade Notes HPLC
Molecular Formula C8 H6 N2 O
Quantity 500 mg
Synonym 1(2H)-Phthalazinone, 1-Hydroxyphthalazine, 1-Oxophthalazine, 1-Phthalazinol, ICX 56225770, NSC 10432, NSC 52567, Phthalazin-1-one, Phthalazinone
InChI Formula InChI=1S/C8H6N2O/c11-8-7-4-2-1-3-6(7)5-9-10-8/h1-5H,(H,10,11)
SMILES O=C1NN=Cc2ccccc12
IUPAC Name 2H-phthalazin-1-one
Molecular Weight (g/mol) 146.15
Formula Weight 146.048
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.