missing translation for 'onlineSavingsMsg'
Learn More

Phthalazine, TRC

Product Code. 30418799
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418799 25 g 25g
30423409 1 g -
30415232 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418799 Supplier Toronto Research Chemicals Supplier No. TRCP38250025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253-52-1
Molecular Formula C8 H6 N2
Molecular Weight (g/mol) 130.15
Synonym 2,3-Benzodiazine, 2,3-Diazanaphthalene, Benzo[d]pyridazine, NSC 62484, NSC 63241, β-Phenodiazine
IUPAC Name phthalazine
SMILES c1ccc2cnncc2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H6N2/c1-2-4-8-6-10-9-5-7(8)3-1/h1-6H
Formula Weight 130.0531
Percent Purity >95%
Chemical Name or Material Phthalazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.