Learn More
Phthalazine, 98%
Description
Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.
Solubility
Soluble in water, methanol, chloroform.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 89°C to 92°C |
| Boiling Point | 317°C |
| Quantity | 5 g |
| Beilstein | 109240 |
| Merck Index | 14,7370 |
| Solubility Information | Soluble in water,methanol,chloroform. |
| Formula Weight | 130.15 |
| Percent Purity | 98% |
| Chemical Name or Material | Phthalazine |
Safety and Handling
- Warning
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.