Learn More
Phthalates Mixture 955 1000 μg/mL in Isooctane, Dr. Ehrenstorfer
Product Code. 30311802
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30311802
Brand: Dr. Ehrenstorfer DREGA09000955IO
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester
CAS 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7, 131-11-3
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL