missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalates Mixture 954 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester |
| CAS | 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7, 131-11-3 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Dibutyl phthalate 100 μg/mL∣Diethyl phthalate 100 μg/mL∣Di-n-octyl phthalate 100 μg/mL∣Phthalic acid, benzylbutyl ester 100 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 100 μg/mL∣Phthalic acid, bis-methyl ester 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?