Learn More
Phthalates Mixture 953 1000 μg/mL in Isooctane, Dr. Ehrenstorfer
Product Code. 30321548
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30321548 1 mL 1mL
1 options
Product Code. 30321548 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000953IO
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Adipic Acid Bis(2-ethylhexyl) Ester∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester
CAS 103-23-1, 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Adipic Acid Bis(2-ethylhexyl) Ester 1000 μg/mL∣Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL